Magnesium sulphamate hydrate ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
39381.30
39381.A1
39381.30EA
80.5
EUR
39381.30
39381.A1
Magnesium sulphamate hydrate ≥99%
Magnesium sulphamate hydrate
Purity: 99%
CAS No.: 23221-99-0
Molecular Formula: Mg(SO3NH2)2.xH2O
Formula Weight: 216.48(anhy)
Physical Form: Powder
MDL No.: MFCD00210632
Formel:
Mg(SO₃NH₂)₂·xH₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00210632 CAS-Nummer: 13770-91-7 EINECS: 237-398-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...