Magnesium molybdenum oxide ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Magnesium molybdate

39376.18 39376.30
39376.18EA 147 EUR
39376.18 39376.30
Magnesium molybdenum oxide ≥99,9% (Metall-Basis)
Magnesium molybdenum oxide

Purity: 99.9% CAS No.: 13767-03-8 Molecular Formula: MgMoO4 Formula Weight: 184.25 Physical Form: -200 Mesh Powder Density: 2.208 Odour: Odourless MDL No.: MFCD00016195

Formel: MgMoO₄
Molekulargewicht: 184.243 g/mol
Dichte: 2.208
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00016195
CAS-Nummer: 13767-03-8
EINECS: 237-376-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR