Yttrium aluminum oxide ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Purity: 99.99%
CAS No.: 12005-21-9
Molecular Formula: Y3Al5O12
Formula Weight: 593.62
Physical Form: 3-12mm Fused Lump
Density: 4.65
Refractive Index: 1.833
Melting Point: 1760°
MDL No.: MFCD00213958
Applications: Can be used as a sputtering target to prepare bubble memory devices.
Formel:
Y₃Al₅O₁₂ Schmelzpunkt: 1760 °C Dichte: 4,65 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00213958 CAS-Nummer: 12005-21-9 EINECS: 234-465-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...