(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer Dimer ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Dichloro(pentamethylcyclopentadienyl)rhodium (III) dimer, [Rh(Cp-)Cl2]2, Dichloro(pentamethylcyclopentadienyl)rhodium(III) dimer, Dichloro(pentamethylcyclopentadienyl)rhodium(III) Dimer

33657.03 33657.MD
33657.03EA 683 EUR
33657.03 33657.MD
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer Dimer ≥99%
(Pentamethylcyclopentadienyl)rhodium(III) dichloride dimer

Purity: 99% CAS No.: 12354-85-7 Molecular Formula: C10H15Cl4Rh2 Formula Weight: 618.08 Physical Form: Powder Appearance: Red Odour: Odourless Melting Point: 300° MDL No.: MFCD00061552

Formel: C₂₀H₃₀Cl₄Rh₂
Molekulargewicht: 618.07 g/mol
Schmelzpunkt: >300 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00061552
CAS-Nummer: 12354-85-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR