Insulating tubes, ceramic
Lieferant: Thermo Fisher Scientific
31475.DR 31476.DR 31476.DT 31479.DR 31479.DW 31492.DR 31475.DT 31475.DW 31485.DR 31485.DW 31487.DR 31487.DW
31475.DREA 35.8 EUR
31475.DR 31476.DR 31476.DT 31479.DR 31479.DW 31492.DR 31475.DT 31475.DW 31485.DR 31485.DW 31487.DR 31487.DW
Insulating tubes, ceramic
Schläuche Transferschlauch
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.