1,10-Phenanthrolin-4,7-diol
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,10-Phenanthroline-4,7-diol
31434.02
31434.04
31434.09
31434.02EA
76.1
EUR
31434.02
31434.04
31434.09
1,10-Phenanthrolin-4,7-diol
1,10-Phenanthrolin-4,7-diol
CAS No.: 3922-40-5
Molecular Formula: C12H8N2O2
Formula Weight: 212.21
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: Yellow
Melting Point:
Formel:
C₁₂H₈N₂O₂ Molekulargewicht: 212.21 g/mol Schmelzpunkt: >300 °C (Zers.) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00004975 CAS-Nummer: 3922-40-5 EINECS: 223-493-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...