Bis(2,4-pentanedionato)tin(IV) dichloride ≥95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Tin(IV) Acetylacetonate Dichloride, Tin(IV) Chloride Bis(2,4-pentanedionate), Acetylacetone Tin(IV) Dichloride Salt
30532.09
30532.18
30532.09EA
104
EUR
30532.09
30532.18
Bis(2,4-pentanedionato)tin(IV) dichloride ≥95%
Bis(2,4-pentanedionato)tin(IV) dichloride
Purity: 95%
CAS No.: 16919-46-3
Molecular Formula: SnCl2(C5H7O2)2
Formula Weight: 387.83
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 199°
UN No.: UN3146
MDL No.: MFCD00015316
Formel:
C₁₀H₁₄Cl₂O₄Sn Schmelzpunkt: 199 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00015316 CAS-Nummer: 16919-46-3 EINECS: 240-972-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...