Potassium tricyanomethanide
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
20702.03
20702.06
20702.03EA
192
EUR
20702.03
20702.06
Potassium tricyanomethanide
Potassium tricyanomethanide
CAS No.: 34171-69-2
Molecular Formula: C4KN3
Formula Weight: 129.16
Physical Form: Crystalline
Appearance: Off-white
UN No.: UN3439
MDL No.: MFCD00058850
Formel:
C₄KN₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00058850 CAS-Nummer: 34171-69-2 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...