Iron(III) meso-tetraphenylporphine chloride
Lieferant: Thermo Fisher Scientific
Synonyme:
Iron(III) Tetraphenylporphyrin chloride, 5,10,15,20-Tetraphenyl-21H,23H-porphine Iron(III) chloride, meso-Tetraphenylporphyrin iron(III) chloride, iron(III) chloride TPP, 21H,23H-Porphine, 5,10,15,20-tetraphenyl-, iron complex, 5,10,15,20-Tetraphenyl-21H,23H-porphine iron(III) chloride
CAS No.: 16456-81-8
Molecular Formula: C44H28ClFeN4
Formula Weight: 704.04
Physical Form: Crystalline
MDL No.: MFCD00012152
Formel:
C₄₄H₂₈ClFeN₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012152 CAS-Nummer: 16456-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...