Diphenyldithiophosphonic acid
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Diphenylphosphinodithioic acid
17758.02
17758.04
17758.09
17758.02EA
56.5
EUR
17758.02
17758.04
17758.09
Diphenyldithiophosphonic acid
Diphenyldithiophosphonic acid
CAS No.: 1015-38-9
Molecular Formula: (C6H5)2P(S)SH
Formula Weight: 250.31
Physical Form: Crystalline
Melting Point: 55-56°
MDL No.: MFCD00014457
Formel:
(C₆H₅)₂P(S)SH Schmelzpunkt: 55…56 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014457 CAS-Nummer: 1015-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...