Molybdenum, platinum coated, Drahtform, Ø 0.457 mm (0.018 in)

Lieferant: Thermo Fisher Scientific

14658.BU 14658.G1 14658.G5
14658.BUEA 60.2 EUR
14658.BU 14658.G1 14658.G5
Molybdenum, platinum coated, Drahtform, Ø 0.457 mm (0.018 in)
Molybdenum, platinum coated

CAS No.: 7439-98-7 Physical Form: Pt thickness 2 micron MDL No.: MFCD00003465

Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003465
CAS-Nummer: 7439-98-7
EINECS: 231-107-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR