Yttrium(III) acetate tetrahydrate ≥99,9%

Lieferant: Thermo Fisher Scientific

14565.30 14565.18
14565.30EA 284 EUR
14565.30 14565.18
Yttrium(III) acetate tetrahydrate ≥99,9%
Yttrium(III) acetate tetrahydrate

Purity: 99.9% CAS No.: 85949-60-6 Molecular Formula: C6H9O6Y.4H2O Formula Weight: 338.10 (266.04anhy) Physical Form: Powder Density: 1.5 Appearance: White Melting Point: dec. Solubility: Soluble in hot water MDL No.: MFCD00013048


Applications: In analytical chemistry

Formel: C₆H₉O₆Y·4H₂O
Schmelzpunkt: Dec.
Dichte: 1.5
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013048
CAS-Nummer: 85949-60-6
EINECS: 245-612-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR