Tris(cyclopentadienyl)yttrium(III) ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Tris(cyclopentadienyl)yttrium
ampouled under argon
14478.02
14478.04
14478.02EA
108
EUR
14478.02
14478.04
Tris(cyclopentadienyl)yttrium(III) ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden)
Tris(cyclopentadienyl)yttrium(III)
Purity: 99.9%
CAS No.: 1294-07-1
Molecular Formula: C15H15Y
Formula Weight: 284.20
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Melting Point: 295°
Boiling Point: 200° subl. (vacuum)
UN No.: UN3181
MDL No.: MFCD00058706
Formel:
C₁₅H₁₅Y Siedepunkt: 200 °C (subl. in vacuum) Schmelzpunkt: 295 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00058706 CAS-Nummer: 1294-07-1 EINECS: 215-069-4 UN-Nummer: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...