Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtgeflecht 80 mesh woven from 0.127mm (0.005in) Type 304
Lieferant: Thermo Fisher Scientific
13477.CH
13477.RR
13477.CHEA
127
EUR
13477.CH
13477.RR
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), Drahtgeflecht 80 mesh woven from 0.127mm (0.005in) Type 304
Stainless steel
Molecular Formula: Fe:Cr:Ni; 70:19:11 wt%
Physical Form: Wire Cloth
MDL No.: MFCD00148499