Holmium(III)-2,4-pentanedionate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
13199.04
13199.09
13199.04EA
166
EUR
13199.04
13199.09
Holmium(III)-2,4-pentanedionate ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Holmium(III)-2,4-pentanedionate
Purity: 99.9%
CAS No.: 14589-33-4
Molecular Formula: C15H21HoO6
Formula Weight: 462.26
Physical Form: Powder
Sensitivity: Hygroscopic
Odour: Odourless
MDL No.: MFCD00050176
Formel:
C₁₅H₂₁HoO₆ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00050176 CAS-Nummer: 14589-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...