Yttrium(III) sulphate hydrate ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
13155.14
13155.22
13155.14EA
121
EUR
13155.14
13155.22
Yttrium(III) sulphate hydrate ≥99,99% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Yttrium(III) sulphate hydrate
Purity: 99.99%
Molecular Formula: Y2(SO4)3.xH2O
Formula Weight: 465.99(anhy)
Physical Form: Crystalline
MDL No.: MFCD00011472
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...