Iron sulphide ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific

Synonyme: Iron pyrite, Iron disulphide

12842.03 12842.09
12842.03EA 62.9 EUR
12842.03 12842.09
Iron sulphide ≥99,9% (Metall-Basis)
Iron sulphide

Purity: 99.9% CAS No.: 12068-85-8 Molecular Formula: FeS2 Formula Weight: 119.98 Physical Form: -100 Mesh Powder Density: 4.87 Odour: Odourless Melting Point: 1171° MDL No.: MFCD00064690

Formel: FeS₂
Schmelzpunkt: 1171 °C
Dichte: 4.87
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00064690
CAS-Nummer: 12068-85-8
EINECS: 235-106-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR