Norbornadiene rhodium(I) chloride dimer (≥44% Rh)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Chloronorbornadiene rhodium(I) dimer, Bis(norbornadiene-mu-chlororhodium), Chloronorbornadiene Rhodium(I) Dimer

10467.02 10467.03
10467.02EA 652 EUR
10467.02 10467.03
Norbornadiene rhodium(I) chloride dimer (≥44% Rh)
Norbornadiene rhodium(I) chloride dimer

CAS No.: 12257-42-0 Molecular Formula: C14H16Cl2Rh2 Formula Weight: 461.01 Physical Form: Powder Odour: Odourless MDL No.: MFCD00198060

Formel: C₁₄H₁₆Cl₂Rh₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00198060
CAS-Nummer: 12257-42-0
EINECS: 235-510-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR