Carbonylhydridotris(triphenylphosphine)rhodium(I) (≥10,0% Rh)
Lieferant: Thermo Fisher Scientific
Synonyme:
Tris(triphenylphosphine)rhodium(I) Carbonyl Hydride, Carbonyltris(triphenylphosphine)rhodium(I) Hydride, Hydridocarbonyltris(triphenylphosphine)rhodium(I)
CAS No.: 17185-29-4
Molecular Formula: C55H46OP3Rh
Formula Weight: 918.80
Physical Form: Powder
Density: 1.33
Sensitivity: Moisture Sensitive
Appearance: Yellow
Odour: Odourless
Melting Point: 138° dec
Solubility: Soluble in hydrocarbons (e.g. benzene and toluene) with dissociation of phosphine ligands
MDL No.: MFCD00151644
Applications: Hydrogenation, hydrosilation, isomerization, carbonylation, hydroformylation, oxidation
Formel:
C₅₅H₄₆OP₃Rh Schmelzpunkt: 138 °C (dec.) Dichte: 1.33 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151644 CAS-Nummer: 17185-29-4 EINECS: 241-230-3 Merck Index: 14,9758 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...