D-Glucoronic acid zur Synthese, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
M-Clarity™ Qualitätsstufe = MQ200
8.43961.0010
8.43961.0025
8.43961.0010EA
119
EUR
8.43961.0010
8.43961.0025
D-Glucoronic acid zur Synthese, Sigma-Aldrich®
D-Glucoronic acid
Formel:
C₆H₁₀O₇ Molekulargewicht: 194.139 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00166981 CAS-Nummer: 6556-12-3 EINECS: 229-486-4 |
Spezifikation Testergebnisse
Solubility | Soluble at 20 °C |
Melting Point | 160...164 °C |
Hill Formula | C6H10O7 |
Molar Mass | 194,14 g/mol |
HS Code | 2932 99 00 |
Storage temperature | 15...25 °C |
Assay (acidimetric) | ≥98% |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...