Separating funnel, pear shaped
Lieferant: SMITH SCIENTIFIC
8S152/250 8S150/250
SMIT8S152/250EA 160 EUR
SMIT8S152/250 SMIT8S150/250
Separating funnel, pear shaped
Trichter Scheidetrichter
Borosilicate glass 3.3, with plastic stopper.
  • Stopcock with glass key
  • Very good temperature resistance
  • Very good chemical resistance