Extraction disk holders, Diskcover-47
Lieferant: Restek
24021 24020
REST24021EA 1553 EUR
REST24021 REST24020
Extraction disk holders, Diskcover-47
Filterhalter
Extraction disk holders are compatible with most vacuum manifold systems that accept ⅛" male luer fittings. Samples can be automatically introduced via ⅛" PTFE tubing or from the Diskcover-47 reservoir.