Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB7]

Lieferant: Tonbo Biosciences
50-0388-T025 50-0388-T100 50-0388-T500
50-0388-T025EA 86.3 EUR
50-0388-T025 50-0388-T100 50-0388-T500
Anti-CD38 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HB7]
Antikörper
The HB7 antibody is specific for human CD38, a 45 kDa type II transmembrane glycoprotein expressed on thymocytes, plasma cells, and monocytes as well as other non-hematopoietic cells. T and B lymphocyte expression is discontinuous and varies based on differentiation state � present on immature cells, low on intermediate stages, and high on activated mature cells. It is found on most CD34+ cells, but not on pluripotent stem cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HB7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR