Anti-CD45RA Mouse Monoclonal Antibody (redFluor™ 710) [clone: HI100]

Lieferant: Tonbo Biosciences
80-0458-T025 80-0458-T100
80-0458-T025EA 101 EUR
80-0458-T025 80-0458-T100
Anti-CD45RA Mouse Monoclonal Antibody (redFluor™ 710) [clone: HI100]
Antikörper
The HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RB, CD45RO and others.

The HI100 antibody is widely used as a marker for human CD45RA expression on naïve and activated T cells, B cells, and monocytes.

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: HI100
Conjugation: redFluor™ 710
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human, Chimpanzee
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR