Anti-GM-CSF Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
143579 143579-50
USBI143579EA 324 EUR
USBI143579 USBI143579-50
Anti-GM-CSF Rabbit Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: ELISA: 0.25-1 ?g/ml

Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR