Anti-GM-CSF Rabbit Polyclonal Antibody
Lieferant: US Biological
143578 143578-100
USBI143578EA 324 EUR
USBI143578 USBI143578-100
Anti-GM-CSF Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-2?g/ml
Type: Primary
Antigen: GM-CSF
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat,