Anti-AURKA Mouse Monoclonal Antibody [clone: 1F8]

Lieferant: Novus Biologicals
NBP2-22118 NBP2-22118SS
NOVUNBP2-22118EA 554 EUR
NOVUNBP2-22118 NOVUNBP2-22118SS
Anti-AURKA Mouse Monoclonal Antibody [clone: 1F8]
Antikörper
Type: Primary
Antigen: AURKA (aurora kinase A)
Clonality: monoclonal
Clone: 1F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR