Anti-HSPA13 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-77456 NBP1-77456SS
NOVUNBP1-77456EA 582 EUR
NOVUNBP1-77456 NOVUNBP1-77456SS
Anti-HSPA13 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal Hsp70 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR