Anti-NBR1 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NBP1-71703 NBP1-71703SS
NOVUNBP1-71703EA 638 EUR
NOVUNBP1-71703 NOVUNBP1-71703SS
Anti-NBR1 Rabbit Polyclonal Antibody
Antikörper
Rabbit Polyclonal NBR1 Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: NBR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR