Anti-IgG Donkey Polyclonal Antibody (CF™ 543)

Lieferant: Biotium
20322 20322-1
20322.EA 359 EUR
20322. 20322-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Antikörper
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 543 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR