Anti-IgG Donkey Polyclonal Antibody (CF™ 350)

Lieferant: Biotium
20275 20275-1
20275.EA 359 EUR
20275. 20275-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 350)
Antikörper
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 350 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR