Anti-IgG Donkey Polyclonal Antibody (CF™ 680)

Lieferant: Biotium
20241 20241-1
20241.EA 368 EUR
20241. 20241-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
Antikörper
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 680 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Guinea pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR