Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Lieferant: Biotium
20130 20130-1
20130.EA 238 EUR
20130. BTIU20130-1
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antikörper
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™633 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™633 (Ex/Em 630/650 nm) produces brighter antibody conjugates than Alexa Fluor® 647, and has unmatched photostability.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF633
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse