Anti-IgG Donkey Polyclonal Antibody (CF™ 647)

Lieferant: Biotium
20046 20046-1
20046.EA 359 EUR
20046. 20046-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 647)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR