Anti-cAMP Protein Kinase Catalytic subunit Rabbit Monoclonal Antibody [clone: EP2102Y]
Lieferant: Abcam
AB76238-100 AB76238-1ML
ABCAAB76238-100EA 669 EUR
ABCAAB76238-100 ABCAAB76238-1ML
Anti-cAMP Protein Kinase Catalytic subunit Rabbit Monoclonal Antibody [clone: EP2102Y]
Antikörper
Rabbit monoclonal [EP2102Y] to cAMP Protein Kinase Catalytic subunit.
Bitte beachten All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: cAMP Protein Kinase Catalytic subunit
Clonality: Monoclonal
Clone: EP2102Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat