Anti-STAR Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB129747-100 ORB129747-200
BIRBORB129747-100EA 0 EUR
BIRBORB129747-100 BIRBORB129747-200
Anti-STAR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STAR (steroidogenic acute regulatory protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR