Anti-AR Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB127595-100 ORB127595-200
BIRBORB127595-100EA 0 EUR
BIRBORB127595-100 BIRBORB127595-200
Anti-AR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AR (androgen receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR