Anti-AR Rabbit polyclonal antibody

Lieferant: Biorbyt
ORB106359-100 ORB106359-20
BIRBORB106359-100EA 0 EUR
BIRBORB106359-100 BIRBORB106359-20
Anti-AR Rabbit polyclonal antibody
Antikörper
  • Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: AR (androgen receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR