Anti-Melamine Rabbit Polyclonal Antibody
Lieferant: Biorbyt
ORB11037-100 ORB11037-200
BIRBORB11037-100EA 0 EUR
BIRBORB11037-100 BIRBORB11037-200
Anti-Melamine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Melamine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen