Anti-AR Rabbit polyclonal antibody

Lieferant: Biorbyt
ORB10128-100 ORB10128-200
BIRBORB10128-100EA 0 EUR
BIRBORB10128-100 BIRBORB10128-200
Anti-AR Rabbit polyclonal antibody
Antikörper
  • ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: AR (androgen receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR