Anti-Melamine Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB10002-100 ORB10002-200
BIRBORB10002-100EA 0 EUR
BIRBORB10002-100 BIRBORB10002-200
Anti-Melamine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Melamine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR