Anti-Melamine Rabbit Polyclonal Antibody

Lieferant: Biorbyt
ORB5636-100 ORB5636-200
BIRBORB5636-100EA 0 EUR
BIRBORB5636-100 BIRBORB5636-200
Anti-Melamine Rabbit Polyclonal Antibody
Antikörper
ELISA: 1:500-1000 (based on 0.5 mg/ml)

Type: Primary
Antigen: Melamine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR