Anti-FL 393 Rabbit polyclonal antibody

Lieferant: Biorbyt
ORB2664-100 ORB2664-200
BIRBORB2664-100EA 0 EUR
BIRBORB2664-100 BIRBORB2664-200
Anti-FL 393 Rabbit polyclonal antibody
Antikörper
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: FL 393
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR