Anti-Ly-6G Rat Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: 1A8]

Lieferant: Tonbo Biosciences
65-1276-U100 65-1276-U025
65-1276-U100EA 196 EUR
65-1276-U100 65-1276-U025
Anti-Ly-6G Rat Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: 1A8]
Antikörper
Type: Primary
Antigen: Ly-6G (Ly-6G)
Clonality: monoclonal
Clone: 1A8
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR