3,3',5,5'-Tetramethylbenzidin (TMB) Membrane Peroxidase Substrate
Lieferant: Rockland Immunochemicals
Synonyme: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
TMBE-100 TMBM-100
ROCKTMBE-100EA 233 EUR
ROCKTMBE-100 ROCKTMBM-100
3,3',5,5'-Tetramethylbenzidin (TMB) Membrane Peroxidase Substrate
3,3',5,5'-Tetramethylbenzidin (TMB)
In the presence of peroxidases, TMB substrates can act as an electron donor for the conversion of peroxides to water.
  • Reaction can be halted with acid to change the TMB to yellow
  • Soluble blue end product read at 370 nm or 655 nm
  • Incubation time varies depending on the assay conditions
TMB ELISA peroxidase substrate (3, 3', 5, 5'-tetramethylbenzidine) is a chromogenic substrate used for antibody reactivity in ELISA experiments.
TMB ELISA peroxidase substrate is designed for use in immunology and cancer research.
Lieferumfang: Supplied as a ready to use clear to very light blue colour liquid.
Formel: C₁₆H₂₀N₂
Molekulargewicht: 240.35 g/mol
Siedepunkt: 402
MDL Number: MFCD00007748
CAS-Nummer: 54827-17-7