Precision tweezers, straight
Lieferant: Roth Carl
K339.1 K341.1 K342.1
ROTHK339.1EA 0 EUR
ROTHK339.1 ROTHK341.1 ROTHK342.1
Precision tweezers, straight
Pinzetten
Magnetic stainless steel (Inox 02 or Inox 08).
  • Extremely fine tips
  • Ideal for high precision laboratory work, e.g. microscopy, biology and microbiology
  • Temperature-resistant up to 400 °C, according to DIN 50914
  • Inox 08 tweezers: Excellent resistance to corrosion, good resistance to salt, autoclavable