Standard conical ground joints sockets and cones
Lieferant: SMITH SCIENTIFIC
8120/50/42 8124/12/21 803/24/29 803/29/32 8124/24/40
SMIT8120/50/42EA 79.5 EUR
SMIT8120/50/42 SMIT8124/12/21 SMIT803/24/29 SMIT803/29/32 SMIT8124/24/40
Standard conical ground joints sockets and cones
Adapter Übergangsstücke mit Kegelschliffen
Made of borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes