Büchner-Trichter
Lieferant: SMITH SCIENTIFIC
34S/30/3 34S/60/3 34S/90/3 34S/120/4 34S/175/4 34S/30/4 34S/40/4 34S/60/4 34S/90/4
SMIT34S/30/3EA 916 EUR
SMIT34S/30/3 SMIT34S/60/3 SMIT34S/90/3 SMIT34S/120/4 SMIT34S/175/4 SMIT34S/30/4 SMIT34S/40/4 SMIT34S/60/4 SMIT34S/90/4
Büchner-Trichter
Trichter Filtertrichter Büchner-Trichter
Funnels, made of, borosilicate glass, with sintered glass disc. For convenient filtration of solutions.
  • No filter papers required
  • Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
  • Excellent temperature resistance