Puromycin Dihydrochlorid
Lieferant: Cayman Chemical
|
Achtung
|
13884-25
13884-50
13884-100
13884-500
CAYM13884-25EA
47
EUR
CAYM13884-25
CAYM13884-50
CAYM13884-100
CAYM13884-500
Puromycin Dihydrochlorid
Puromycin Dihydrochlorid
Formel:
C₂₂H₂₉N₇O₅·2HCl Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00012691 CAS-Nummer: 58-58-2 EINECS: 200-387-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...