Xylenolorange Ditriumsalz für die Spektrophotometrie, Supelco®
Lieferant: Merck
Synonyme:
3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulphonephthalein disodium salt
M-Clarity™ quality level = MQ100
52097-1G
52097-5G
52097-1GEA
52.9
EUR
52097-1G
52097-5G
Xylenolorange Ditriumsalz für die Spektrophotometrie, Supelco®
Xylenolorange Ditriumsalz
Reagent for spectrophotometric determination of metal ions
Formel:
C₃₁H₃₀N₂Na₂O₁₃S |
MDL Number:
MFCD00014580 CAS-Nummer: 1202864-39-8 EINECS: 216-553-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...